Dichloroacetic acid (DCA), GPR RECTAPUR®
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
DCA
87085.290EA
114
GBP
87085.290
87085.230
Dichloroacetic acid (DCA), GPR RECTAPUR®
Dichloroacetic acid (DCA)
Formula:
C₂H₂Cl₂O₂ MW: 128.94 g/mol Boiling Pt: 194 °C (1013 hPa) Melting Pt: ∼–4 °C Density: 1.563 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004223 CAS Number: 79-43-6 EINECS: 201-207-0 UN: 1764 ADR: 8,II |
Specification Test Results
Appearance | Clear liquid |
Assay (GC) | ≥98.0 % |
Density (d20°C/ 4°C) | 1.560 - 1.570 |
Identity (IR) | passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...