Fluoride Activity Standard Solution for Fluoride Electrode, Supelco®
Supplier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
443174-475MLEA 147 GBP
443174-475ML
Fluoride Activity Standard Solution for Fluoride Electrode, Supelco®
Standards Anionic Standards Fluoride Standards
Activity standard solution consists of sodium fluoride solution. 100 ppm as fluoride.
It may be used as calibration standard in determination of fluoride in bones using fluoride electrode.
100 ppm = 100 mg/L (as F).