RABS/isolator gloves, high grip, BioClean™ Sterile Nitrile GHG
Supplier: Nitritex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
NITRGHG33NIT59EA 0 GBP
NITRGHG33NIT59
RABS/isolator gloves, high grip, BioClean™ Sterile Nitrile GHG
These sterile RABS and GHG isolator gloves are manufactured from nitrile with incredibly low levels of particles and excellent ESD properties.
  • Ultra-clean surface ensures product protection
  • 100% inspected and air leak tested
  • Suiatble for autoclaving
  • Can be sanitised by VHP or IPA
  • Gamma irradiated sterilisation; sterility Assurance Level (SAL) of 10⁻⁶
  • High grip
Designed for precision work when increased grip is required, the GHG series of high grip gloves are available in a range of port sizes.
Compatible with ISO Class 4 & EU GMP Grade A cleanrooms.
Certifications: EN 388:2016 (4101X), EN ISO 374-1 Type A (AJKNOT), EN ISO 374-5 VIRUS, EN 421; Cat. III
Food contact approved
Tested against permeation standard ASTM D6978-05 for cytotoxic drugs.
Packaging: Triple bagged: One piece per sealed inner PE bag; one inner bag per sealed second inner PE bag; one second inner bag per sealed outer PE bag; 20 outer bags per lined inner white Correx polyethylene box (20 pieces).