An ideal affinity purification matrix to deplete human albumin and IgG from plasma and serum samples for down stream processing.
The Hi-Bind™ albumin-IgG depletion beads are prepared by covalent conjugation to cross-linked (4%) Agarose beads by a proprietary method. These depletion beads can be used as an affinity purification matrix to remove human albumin and IgG from plasma and serum samples for subsequent ELISA, immunoassays and other downstream analyses such as gel electrophoresis, functional assays and more.
Caution: For research use only. Not for use in humans.