DNA Clean-Up, MicroSpin Cycle-Pure Kits, peqGOLD

Supplier: VWR Peqlab
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

peqGOLD
13-6293-00EA 7.8 GBP
13-6293-00 13-6293-01 13-6293-02
DNA Clean-Up, MicroSpin Cycle-Pure Kits, peqGOLD
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Clean up DNA in low elution volume (10 to 15 µl) from PCR reactions using spin columns.

  • Starting amount up to 10 µg DNA
  • DNA recovered >90% recovery, 70 bp to 20 kb

Modified from the peqGOLD Cycle-Pure Kit.

Delivery information: Each kit comprises of peqGOLD MicroSpin DNA columns, 2 ml collection tubes, CP buffer, DNA wash buffer and Elution buffer.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR