Lard oil, analytical standard, Supelco®
Supplier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
47115-UEA 80.9 GBP
47115-U
Lard oil, analytical standard, Supelco®
Standards Food Standards
This reference material is offered as a qualitative standard. Use the chromatographic fingerprint of our reference material to check for the presence of lard oil in your sample.
Storage temperature: 2 to 8 °C
Certifications: A Certificate of Composition which includes a chromatogram is shipped with each product.