F.A.M.E. Mix GLC-100, certified reference material, TraceCERT®, Supelco®
Supplier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TraceCERT®
1899-1AMPEA 67.9 GBP
1899-1AMP
F.A.M.E. Mix GLC-100, certified reference material, TraceCERT®, Supelco®
Standards Food Standards
This quantitative mix is useful for determining relative retention times and approximating response factors
Methyl arachidate 20 wt. %; Methyl behenate 20 wt. %; Methyl heneicosanoate 20 wt. %; Methyl nonadecanoate; Methyl stearate 20 wt. %