Pesticide reference standard, Bentazon, PESTANAL®, analytical standard, Supelco®
Supplier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PESTANAL®
32052-250MGEA 49.9 GBP
32052-250MG
Pesticide reference standard, Bentazon, PESTANAL®, analytical standard, Supelco®
Standards Organic Standards
Bentazon is a commonly used pesticide which effectively controls weed in paddy fields. It has been used as analytical standard in liquid chromatograph with a photodiode array (PDA) detector coupled in series with a mass spectroscopy (MS) detector equipped with an atmospheric pressure electrospray ionization source (ESI) for the determination of bentazon and its transformation products.