Alkaline earth metal mix for ICP, 5 elements, TraceCERT®, Supelco®
Supplier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TraceCERT®
67288-100MLEA 113 GBP
67288-100ML
Alkaline earth metal mix for ICP, 5 elements, TraceCERT®, Supelco®
Standards Cationic Standards Multi-Element Standards
100 mg/L each: Be, Mg, Ca, Sr and Ba in 2% nitric acid
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34).