This triple quadrupole calibration solution, extended mass range, is a mixture of 14 highly pure, ionizable components designed for both positive and negative ionization calibration of triple stage quadrupole instruments.
- Improved mass accuracy
- Highly validated
- Ready to use
- Stable - store at 2 to 8 °C for 18 months
- Each lot is quality controlled with strict specifications
The extended range masses improve performance of the triple quadrupole mass spectrometers. This formulation contains 14 compounds, that are specifically optimized for use with TSQ Endura™ Series and TSQ Quantiva™ series instruments. The calibration solution is also compatible with the TSQ Quantum™ series, TSQ Discovery™ series, TSQ Quantum Ultra™ series, TSQ Quantum Access™ series, and TSQ Vantage™ series instruments.
Certifications: Manufactured at an ISO 9001 facility.
Packaging: The stable solution is provided in a leak-proof, high-purity PTFE bottle.