Wipes, pre-saturated, Wip'Anios Excel
Supplier: Anios
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
DMDA2446655Y6EA 140 GBP
DMDA2446655Y6
Wipes, pre-saturated, Wip'Anios Excel
Wipes Disposable Wipes and Towels Presaturated Wipes
Wipes for the treatment of surfaces and medical devices.
  • 100% biodegradable viscose wipe
  • Formulated without perfume, nor alcohol
  • Broad virucidal spectrum
  • Microbial efficiency active from 3 minutes
Type-4 biocide: Can be used on surfaces in contact with food.
Packaging: Packed in a re-sealable pouch (100 wipes per pouch).
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904