Bottle carrier
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
212-0702EA 74.1 GBP
VWRI212-0702
Bottle carrier
Made from 304 grade stainless steel, with a wire mesh bottom for increased stability.
  • Sturdy
  • Handles fold down for easier storage
  • Autoclavable
These safety wire bottle carriers will securely transport two ‘Merck’/Winchester style bottles up to 135 mm tall and 170 mm diameter (typically 2,5 litre capacity).