Palladium(II) iodide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

SIAL464678-2GEA 205 GBP
SIAL464678-2G
Palladium(II) iodide, Sigma-Aldrich®
Palladium (II) iodide
Formula: I₂Pd
MW: 360.23 g/mol
Density: 6.003 g/cm³ (18 °C)
MDL Number: MFCD00011171
CAS Number: 7790-38-7
EINECS: 232-203-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR