United Kingdom
Order Entry
Ammonium sulphamate ACS
 
undefined
Ammonium sulphamate ACS

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Melting point:
    132.0 to 134.0°C
  • Residue after ignition:
    ≤0.10 %
  • Titration:
    ≥98.0 %
  • Heavy metals (as Pb):
    ≤5 ppm
  • Insoluble matter:
    ≤0.02 %

Specification Test Results

Related Information

Product Information

  • Catalog #
    Supplier #
    Pack type
    Pk
    Availability
    Price
  • ACRO423390050
    423390050
    Glass bottle for solids
    5 g
       
    Log in for availability
    1 * 5 g
    1 * 5 g - £22.30
    £22.30
    1 * 5 g
       £22.30

  • ACRO423391000
    423391000
    Plastic bottle
    100 g
       
    Log in for availability
    1 * 100 g
    1 * 100 g - £26.80
    £26.80
    1 * 100 g
       £26.80

  • ACRO423395000
    423395000
    Plastic bottle
    500 g
       
    Log in for availability
    1 * 500 g
    1 * 500 g - £82.60
    £82.60
    1 * 500 g
       £82.60

  • ACRO423390025
    423390025
    Plastic bottle
    2,5 kg
       
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - £263.00
    £263.00
    1 * 2,5 kg
       £263.00