Carbon disulphide, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
M-Clarity™ quality level = MQ100
SIAL335266-1LEA
180
GBP
SIAL335266-1L
SIAL335266-100ML
Carbon disulphide, Sigma-Aldrich®
Carbon disulphide
Formula:
CS₂ MW: 76.14 g/mol Boiling Pt: 46.2 °C (1013 hPa) Melting Pt: –111.6 °C Density: 1.2632 g/cm³ (20 °C) Flash Pt: –30 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011321 CAS Number: 75-15-0 EINECS: 200-843-6 UN: 1131 ADR: 3,I Merck Index: 12,01859 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...