Tin(IV) chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Stannic chloride, Tin tetrachloride
CAS Number:7646-78-8
:
Formula:Cl₄Sn
MDL Number:MFCD00011242
Boiling Pt:114
Melting Pt:–33 °C
Density:2.22 g/cm³ (20 °C)
Molecular Weight:260.52 g/mol
EINECS:231-588-9
UN:1827
ADR:8 II
Merck Index:12,08929
:M-Clarity™ quality level = MQ200
:GHS05
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications