Magnesium hydroxide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ200

SIALM5421-1KGEA 206 GBP
SIALM5421-1KG
Magnesium hydroxide, Sigma-Aldrich®
Magnesium hydroxide
Formula: Mg(OH)₂
MW: 58.32 g/mol
Density: 2.3261…2.3631 g/cm³ (20 °C)
MDL Number: MFCD00011104
CAS Number: 1309-42-8
EINECS: 215-170-3
Merck Index: 13,05693

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR