Ammonium hydrogen difluoride
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Ammonium bifluoride
, Ammonium hydrogen fluoride
1.01160.5000EA
179
GBP
1.01160.5000
Ammonium hydrogen difluoride
Ammonium hydrogen difluoride
Formula:
NH₄HF₂ MW: 57.04 g/mol Melting Pt: 127 °C Density: 0.8 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00012018 CAS Number: 1341-49-7 EINECS: 215-676-4 UN: 1727 ADR: 8,II Merck Index: 13,00498 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...