3',5'-Dihydroxyacetophenone
Supplier: Molekula
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-(3,5-Dihydroxyphenyl)ethanone
, 1-(3,5-Dihydroxyphenyl)ethan-1-one
MOLEM62767659EA
0
GBP
MOLEM62767659
3',5'-Dihydroxyacetophenone
3',5'-Dihydroxyacetophenone
Formula:
C₈H₈O₃ MW: 152.15 g/mol Boiling Pt: 296 °C (1013 hPa) Melting Pt: 148 °C |
MDL Number:
MFCD00002290 CAS Number: 51863-60-6 EINECS: 257-480-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...