6-Chloro-m-cresol
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
1-Chloro-2-hydroxy-4-methylbenzene
, 2-Chloro-5-methylphenol
8.18799.0025EA
0
GBP
8.18799.0025
6-Chloro-m-cresol
6-Chloro-m-cresol
Formula:
C₇H₇ClO MW: 142.58 g/mol Boiling Pt: 196 °C (1013 hPa) Melting Pt: 43…46 °C Density: 1.215 g/cm³ (20 °C) |
MDL Number:
MFCD00002175 CAS Number: 615-74-7 EINECS: 210-444-9 UN: 3437 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...