4-Chloro-2,3,5,6-tetrafluorobenzoyl chloride ≥98%
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
L20167.14EA
230
GBP
L20167.14
4-Chloro-2,3,5,6-tetrafluorobenzoyl chloride ≥98%
4-Chloro-2,3,5,6-tetrafluorobenzoyl chloride
Formula:
C₇Cl₂F₄O MW: 246.98 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03001172 CAS Number: 145572-10-7 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...