4-Amino-5-chloro-2-methoxybenzoic acid ≥98%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
4-Amino-5-chloro-o-anisic acid
B24281.14EA
64.9
GBP
B24281.14
B24281.06
4-Amino-5-chloro-2-methoxybenzoic acid ≥98%
4-Amino-5-chloro-2-methoxybenzoic acid
Formula:
C₈H₈ClNO₃ MW: 201.61 g/mol Boiling Pt: 354 °C (1013 hPa) Melting Pt: 205…208 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00038794 CAS Number: 7206-70-4 EINECS: 230-582-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...