9-Octadecynoic acid ≥98%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
L01294.03EA
29.5
GBP
L01294.03
L01294.06
9-Octadecynoic acid ≥98%
9-Octadecynoic acid
Formula:
C₁₈H₃₂O₂ MW: 280.45 g/mol Boiling Pt: 189…190 °C (2 mmHg) Melting Pt: 43…47 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014386 CAS Number: 506-24-1 EINECS: 208-030-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...