UV glow cream
Supplier: Deb Stoko
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EVONUVC200MLEA 307 GBP
EVONUVC200ML
UV glow cream
This cream contains thousands of minute plastic particles which act as "artificial germs" to reveal hygiene deficiencies.
  • Particles glow under ultraviolet light to clearly show the areas the user missed when washing or disinfecting
  • Increases hygiene by ensuring that all germs are eliminated
Ideal for the healthcare industry.