1-Bromo-3,3-dimethylbutanone
Supplier: Molekula
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
1-Bromo-3,3-dimethyl-2-butanone
, 1-Bromopinacolone
MOLE34413636-25GEA
0
GBP
MOLE34413636-25G
1-Bromo-3,3-dimethylbutanone
1-Bromo-3,3-dimethylbutanone
Formula:
C₆H₁₁BrO MW: 179.06 g/mol Boiling Pt: 47…49 °C (1 mmHg) Melting Pt: 76 °C Density: 1.3508 g/cm³ (0 °C) Storage Temperature: Freezer |
MDL Number:
MFCD00000206 CAS Number: 5469-26-1 EINECS: 226-794-0 UN: 2810 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...