Glycerine ≥99%, Electran for molecular biology

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Propane-1,2,3-triolGlycerol1,2,3-Propanetriol

444485BEA 0 GBP
444485B
Glycerine ≥99%, Electran for molecular biology
Glycerine
Formula: HOCH₂CH(OH)CH₂OH
MW: 92.09 g/mol
Boiling Pt: 290 °C (1013 hPa)
Melting Pt: 18.6 °C
Density: 1.26 g/cm³ (20 °C)
Flash Pt: 177 °C (open cup)
Storage Temperature: Ambient
MDL Number: MFCD00004722
CAS Number: 56-81-5
EINECS: 200-289-5
Merck Index: 13,04497
https://uk.vwr.com/stibo/web/std.lang.all/47/74/734774.jpg

Order Now

Specification Test Results

Assay Min. 99.00 %
Appearance Clear viscous liquid
Identification Passes test
DNases Not detected
RNases Not detected
Proteases Not detected
Density (20/4) 1.2560 - 1.2610
Fatty acids 0.02 %
Organic bound chlorine Max. 0.0005 %
pH (5 mol/l) 5.50 - 8.00
Water Max. 0.5 %
Absorbance (260 nm) (0.5 mol/l) Max. 0.070
Absorbance (280 nm) (0,5 mol/l) Max. 0.0200
Heavy metals (as Pb) Max. 0.0005 %
SO4 (Sulphate) Max. 0.001 %
As (Arsenic) Max. 0.0001 %
Cl (Chloride) Max. 0.0001 %
Fe (Iron) Max. 0.0005 %
Pb (Lead) Max. 0.0001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR