Ethanol 93.8-94.7%, GPR RECTAPUR®

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


Danger

Synonyms: Ethyl alcohol

28719DWDPEA 0 GBP
28719DWDP
Ethanol 93.8-94.7%, GPR RECTAPUR®
Ethanol

Caution: WARNING : this alcohol is regulated. Call us.
For duty paid orders in the UK please add DP to the product code.

Formula: C₂H₅OH
MW: 46.07 g/mol
Melting Pt: –114 °C
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3,II

Order Now

Specification Test Results

Assay 93.8 - 94.7 %
Acidity or alkalinity Passes test
IR Spectrum Passes test
Reducing substances Passes test
Volatile impurities Passes test
Aldehydes (as CH3CHO) Max. 0.001 %
Heavy metals + Zn (as Pb) Max. 0.0002 %
Non-volatile matter Max. 0.002 %
Fe (Iron) Max. 0.0001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR