Dimethyl succinate
Supplier: EHRENSTORFER GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Succinic acid dimethyl ester
, 1,4-Butanedioic acid dimethyl ester
, DBE-4 dibasic ester
EHERC16985500EA
94.4
GBP
EHERC16985500
Dimethyl succinate
Dimethyl succinate
Formula:
C₆H₁₀O₄ MW: 146.14 g/mol Boiling Pt: 200 °C (1013 hPa) Melting Pt: 18…19 °C Density: 1,117 g/cm³ (20 °C) Storage Temperature: Ambient |
CAS Number:
106-65-0 EINECS: 203-419-9 Merck Index: 12,09038 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...