3-Bromo-1-propanol for synthesis, Sigma-Aldrich®
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
3-Bromopropan-1-ol
, 1-Bromo-3-hydroxypropane
M-Clarity™ quality level = MQ200
8.20218.0025EA
246
GBP
8.20218.2500
8.20218.9999
8.20218.0025
3-Bromo-1-propanol for synthesis, Sigma-Aldrich®
3-Bromo-1-propanol
Formula:
BrH₂CCH₂CH₂OH MW: 138.99 g/mol Boiling Pt: 145 °C (1013 hPa) Density: 1.6 g/cm³ (20 °C) Flash Pt: 65 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00002942 CAS Number: 627-18-9 EINECS: 210-986-6 |
Specification Test Results
Solubility | 167 g/l (20 °C) |
Molar Mass | 138.99 g/mol |
Boiling Point | 62 °C (7 hPa) |
Vapor Pressure | 0.13 hPa (20 °C) |
Flash Point | 65 °C |
Refractive Index | 1.4858 (20 °C, 589 nm) |
Density | 1.60 g/cm3 (20 °C) |
pH | (10 g/l, H2O) neutral |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...