L(+)-Potassium hydrogen tartrate
Supplier: LGC Standards PROMOCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Potassium hydrogen L-tartrate
, Potassium acid tartrate
, L(+)-Tartaric acid potassium salt
, Potassium Bitartrate
, (R,R)-(+)-Tartaric acid potassium salt
, Potassium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate
PROONIST-188EA
1420
GBP
PROONIST-188
L(+)-Potassium hydrogen tartrate
L(+)-Potassium hydrogen tartrate
Formula:
KOOC(CHOH)₂COOH MW: 188.18 g/mol Melting Pt: 250 °C Density: 1.943 g/cm³ (20 °C) |
MDL Number:
MFCD00065392 CAS Number: 868-14-4 EINECS: 212-769-1 Merck Index: 12,07776 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...