Magnesium chloride hexahydrate
Supplier: Roth Carl
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ROTHA537.2EA
161.61
GBP
ROTHA537.2
ROTHA537.1
ROTHA537.4
ROTHA537.3
Magnesium chloride hexahydrate
Magnesium chloride hexahydrate
Formula:
MgCl₂·6H₂O MW: 203.3 g/mol Boiling Pt: 1412 °C (1013 hPa) Melting Pt: 117 °C Density: 1.57 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149781 CAS Number: 7791-18-6 EINECS: 232-094-6 Merck Index: 13,05684 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...