Magnesium chloride hexahydrate 99.0-102.0%, Electran for molecular biology
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
436992SEA
0
GBP
436992S
436994U
Magnesium chloride hexahydrate 99.0-102.0%, Electran for molecular biology
Magnesium chloride hexahydrate
Formula:
MgCl₂·6H₂O MW: 203.3 g/mol Boiling Pt: 1412 °C (1013 hPa) Melting Pt: 117 °C Density: 1.57 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149781 CAS Number: 7791-18-6 EINECS: 232-094-6 Merck Index: 13,05684 |
Specification Test Results
Appearance (colour) | almost white |
Appearance (description) | fine-crystalline powder |
Assay (complexometric) | 99.0 - 102.0 % |
pH-value (5 %; water) | 5.0 - 6.5 |
Heavy metals (as Pb) | Max. 0.0005 % |
Fe (Iron) | Max. 0.0005 % |
DNases (Exo- and endonucleases) | non detectable |
RNases | non detectable |
Proteases | non detectable |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...