Nitric acid 69%, AnalaR NORMAPUR® analytical reagent
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
101686EEA
0
GBP
101686E
Nitric acid 69%, AnalaR NORMAPUR® analytical reagent
Nitric acid
Formula:
HNO₃ MW: 63.01 g/mol Boiling Pt: 120.5 °C (1013 hPa) Melting Pt: –42 °C Density: 1.4 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011349 EINECS: 231-714-2 UN: 2031 ADR: 8 (5.1),II Merck Index: 13,06608 |
Specification Test Results
Assay | 68.5 - 69.5 % |
Cl (Chloride) | Max. 0.00005 % |
PO4 (Phosphate) | Max. 0.00005 % |
SO4 (Sulphate) | Max. 0.00005 % |
As + Sb (as As) | Max. 0.000001 % |
Al (Aluminium) | Max. 0.000005 % |
Au (Gold) | Max. 0.00001 % |
B (Boron) | Max. 0.000005 % |
Ba (Barium) | Max. 0.000005 % |
Be (Beryllium) | Max. 0.000002 % |
Bi (Bismuth) | Max. 0.00001 % |
Ca (Calcium) | Max. 0.000010 % |
Cd (Cadmium) | Max. 0.000002 % |
Co (Cobalt) | Max. 0.000001 % |
Cr (Chromium) | Max. 0.000002 % |
Cu (Copper) | Max. 0.000002 % |
Fe (Iron) | Max. 0.00002 % |
Ga (Gallium) | Max. 0.000002 % |
Ge (Germanium) | Max. 0.00001 % |
In (Indium) | Max. 0.000002 % |
K (Potassium) | Max. 0.00001 % |
Li (Lithium) | Max. 0.000002 % |
Mg (Magnesium) | Max. 0.000010 % |
Mn (Manganese) | Max. 0.000001 % |
Mo (Molybdenum) | Max. 0.000002 % |
Na (Sodium) | Max. 0.00003 % |
Ni (Nickel) | Max. 0.000002 % |
Pb (Lead) | Max. 0.000005 % |
Pt (Platinum) | Max. 0.00002 % |
Sr (Strontium) | Max. 0.000002 % |
Ti (Titanium) | Max. 0.00001 % |
Tl (Thallium) | Max. 0.000005 % |
V (Vanadium) | Max. 0.000005 % |
Zn (Zinc) | Max. 0.00001 % |
Zr (Zirconium) | Max. 0.00001 % |
Residue on ignition | Max. 0.0005 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...