Sodium hydroxide 5 mol/l (5 N) in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®

Supplier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Caustic soda

4690.EA 0 GBP
4690.
Sodium hydroxide 5 mol/l (5 N) in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: 109 °C (1013 hPa)
Density: ≤1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Titer (mol/l) 4.990-5.010

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR