Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrobromic acid 47%, ARISTAR® for trace analysis

Supplier: VWR Chemicals
Danger
Synonyms: Anhydrous hydrobromic acidHydrogen bromide



Formula: HBr
MW: 80.91 g/mol
Density: 1,47 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011323
CAS Number: 10035-10-6
EINECS: 233-113-0
UN: 1788
ADR: 8,II
Specification Test Results
Assay (acidimetric) Min. 47.0 %
I (Iodide) Max. 10 ppm
Phosphate, Phosphite (as PO4) Max. 0.05 ppm
Sulfate and sulphite (as SO4) Max. 5 ppm
Ag (Silver) Max. 0.002 ppm
Al (Aluminium) Max. 0.010 ppm
As (Arsenic) Max. 0.005 ppm
Au (Gold) Max. 0.005 ppm
Ba (Barium) Max. 0.005 ppm
Be (Beryllium) Max. 0.001 ppm
Bi (Bismuth) Max. 0.005 ppm
Ca (Calcium) Max. 0.050 ppm
Cd (Cadmium) Max. 0.002 ppm
Co (Cobalt) Max. 0.001 ppm
Cr (Chromium) Max. 0.001 ppm
Cu (Copper) Max. 0.001 ppm
Fe (Iron) Max. 0.010 ppm
Ga (Gallium) Max. 0.005 ppm
Hg (Mercury) Max. 0.005 ppm
In (Indium) Max. 0.001 ppm
K (Potassium) Max. 0.020 ppm
Li (Lithium) Max. 0.001 ppm
Mg (Magnesium) Max. 0.010 ppm
Mn (Manganese) Max. 0.001 ppm
Mo (Molybdenum) Max. 0.001 ppm
Ni (Nickel) Max. 0.002 ppm
Pb (Lead) Max. 0.005 ppm
Sn (Tin) Max. 0.005 ppm
Sr (Strontium) Max. 0.001 ppm
Ti (Titanium) Max. 0.001 ppm
Tl (Thallium) Max. 0.002 ppm
V (Vanadium) Max. 0.001 ppm
Zn (Zinc) Max. 0.005 ppm
Zr (Zirconium) Max. 0.001 ppm
Evaporation residue Max. 2 ppm
Zr (Zirconium) Max 0.001 ppm

Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle SAFEBREAK 500 ml 450073U
Get a discount if you order 6 or more. Login in now for details.
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty