Polyethylene glycol 600 for synthesis, Sigma-Aldrich®
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
PEG 600
M-Clarity™ quality level = MQ200
8.07486.1000EA
48
GBP
8.07486.1000
8.07486.9999
8.07486.5001
Polyethylene glycol 600 for synthesis, Sigma-Aldrich®
Polyethylene glycol 600
Formula:
H(OCH₂CH₂)nOH MW: 600 g/mol Boiling Pt: ∼250 °C (1013 hPa) Melting Pt: 17…22 °C Density: 1.13 g/cm³ (20 °C) Flash Pt: 270 °C Storage Temperature: Ambient |
MDL Number:
MFCD00081839 CAS Number: 25322-68-3 EINECS: 500-038-2 |
Specification Test Results
Solubility | (20 °C) soluble |
Melting Point | 17 - 22 °C |
Molar Mass | 8000 g/mol |
Vapor Pressure |
|
Flash Point | 260 °C |
Density | 1.13 g/cm3 (20 °C) |
pH | 5 - 7 (100 g/l, H2O, 20 °C) |
Viscosity kinematic | 14 - 17 mm2/s (20 °C) (50% solution) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...