Silver ICP standard solution, 1,000 mg/l Ag in dil. nitric acid
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BAKR5779.0100EA 162 GBP
BAKR5779.0100
Silver ICP standard solution, 1,000 mg/l Ag in dil. nitric acid
For metal trace analysis in AAS or ICP
Ask for your “customized multi-element standard solution” with your exact composition using the request form
Ask for a quotation
12/31/2022