Chloroform-[D₁] (99.8% D) +1% v/v TMS for NMR spectroscopy

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Chloroform-DTrichloromethane-D1Chloroform-[²H₁]

36470.36EA 0 GBP
36470.36 36470.14 36470.22
Chloroform-[D₁] (99.8% D) +1% v/v TMS for NMR spectroscopy
Chloroform-[D1]
Formula: CDCl₃
MW: 120.38 g/mol
Boiling Pt: 62 °C
Melting Pt: –64 °C
Density: 1.50
Storage Temperature: Refrigerator
MDL Number: MFCD00000827
CAS Number: 865-49-6
EINECS: 212-742-4
UN: 1888
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR