Petroleum spirit, 100 - 120 °C, EMSURE® Reag. Ph. Eur. for analysis, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Petroleum etherGasolinePetrol

M-Clarity™ quality level = MQ300

1.01781.1000EA 51.4 GBP
1.01781.1000
Petroleum spirit, 100 - 120 °C, EMSURE® Reag. Ph. Eur. for analysis, Supelco®
Petroleum spirit
Boiling Pt: 100…120 °C (1013 hPa)
Density: 0.7 g/cm³ (20 °C)
Flash Pt: –10 °C
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR