Pterostilbene ≥98% (by HPLC)
Supplier: Enzo Life SciencesTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(E)-4-(3,5-Dimethoxystyryl)phenol
CYP1A1 inhibitor
Cell permeable natural methoxylated analog of resveratrol (Prod. No. BML-FR104). Antioxidant, antiproliferative, anti-inflammatory, anti-hyperglycemic and anti-diabetic agent. Induces apoptosis. Induces Autophagy. Very potent inhibitor of cytochrome P450 1A1 (CYP1A1) with Ki=0.57µM.Inhibits activator protein 1 (AP-1) and NF-kappaB activation. Moderately inhibits COX-1 and COX-2 (IC50=19.8µM and 83.9µM).
Synthetic.
Formula:
C₁₆H₁₆O₃ MW: 256.2964 g/mol |
MDL Number:
MFCD00238710 CAS Number: 537-42-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...