Alternariol monomethyl ether ≥98%
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3,7-Dihydroxy-9-methoxy-1-methyl-6H-dibenzo[b,d]pyran-6-one
, Alternariol-9-methyl ether
Topoisomerase IIalpha inhibitor
Mycotoxin contaminant of fruit and cereal products. Exhibits antifungal and phytotoxic activity. Has also been shown to possess DNA strand-breaking activity and inhibits the activity of topoisomerase IIα in mammalian cell models.
Isolated from Alternaria sp.
Formula:
C₁₅H₁₂O₅ MW: 272.253 g/mol |
MDL Number:
MFCD00058532 CAS Number: 26894-49-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...