Sample collection cart
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
216-8000EA 0 GBP
VWRI216-8000
Sample collection cart
Stable, secure, transport of collection supplies. This cart provides a stable, mobile centre for blood collection. Designed to hold a Droplet™ blood collection station.
  • Handle for pulling the unit
  • Five wheel, weighted base for stability
  • Designated tray area dimensions: 429×265×172 mm
Contruction: Made of durable stainless steel and aluminium, painted surfaces incorporate Microban® protection.