Surgical mask, tie-on, type IIR, with Visor
Supplier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ANSM85006EA 54.7 GBP
ANSM85006
Surgical mask, tie-on, type IIR, with Visor
This speciality mask with visor is designed to prevent transmission of disease from a health care worker to a patient. (e.g. spit, mucous).
  • Panoramic visor: ideal for situations with high risk of exposure to blood or other body fluids
  • SOFTSEAL™ : The moulded nose bridge with soft foam nose cushion provides the wearer an easy, comfortable, dependable fit
  • SIDEPROTECT™ for reduced leakage and enhanced filtration capacity
  • EASYBREATH™ reduces contact with mouth and makes breathing easier
  • Shell resists collapsing, even in heat and humidity, so the masks holds its shape longer
Protects healthcare staff and patients from infectious diseases. They may also be used to help reduce the risk of splashes or sprays of blood, body fluids, secretions and excretions from reaching the wearer’s mouth and nose.