Sodium chloride 0,9%, Freeflex®

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


Freeflex®
FRES2295121EEA 107 GBP
FRES2295121E FRES2295172E FRESB248941
Sodium chloride 0,9%, Freeflex®
Sodium chloride

These injection solutions are suitable for intravenous infusion.


Packaging: In Freeflex® bag (a flexible bag with self-sealing ports and made of multilayer polyolefin film that is PVC-free, plasticiser-free, latex-free and non-DEHP).

Formula: NaCl
MW: 58.44 g/mol
MDL Number: MFCD00003477
CAS Number: 7647-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR