Whole blood, multiple sclerosis
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HCMSPB000F-2-MSEA 0 GBP
HCMSPB000F-2-MS
Whole blood, multiple sclerosis
Whole blood used for researching the causes, diagnosis, supportive care and treatment of multiple sclerosis (MS). Collected from IRB-consented donors diagnosed with multiple sclerosis.
No pre-screening is done on fresh products, but donors are tested on day of collection for Hepatitis B, Hepatitis C, and HIV 1/2, HTLV-I/II, HIV-1/HCV/HBV Nucleic Acid, WNV Nucleic Acid, Trypanasoma cruzi and Syphilis.
Storage: Use immediately upon receipt.
Caution: Intended for research use only and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.