Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Photo tags

Supplier: Brady
These photo tags can be customised by writing on special instructions, hazards or warnings. They can also have an area where a relevant employee photo can be included, giving a more personal message.

  • Made from heavy duty polyester
  • Employee photos for easy identification and greater impact

Colour: Black and Red on white

Thickness: 0,249 mm

Certifications: OSHA 1910,145(f).

Packaging: 10 tags with nylon fasteners.


 

ORDER

Expand All / Collapse All
Description Dimensions Colour Pk VWR Catalog Number Quantity
Photo tags 76,2×146,05 mm Black/red/white 10 BRDY065500
Self-laminating photo tags 76,2×146,05 mm Black/red/white 10 BRDY096222
Self-laminating photo tags 76,2×177,80 mm Black/red/white 10 BRDY065501
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping