Would you like to log in again?
2D spin-out swabs
Supplier: VWR - Omega Bio-Tek
The 2D spin-out swabs provide a complete solution for the collection and tacking of swabs throughout the DNA extraction process when processed in a high throughput format. The 2D spin-out swabs integrated with the E-Z 96® spin-out device offers a fast, convenient way for sample traceability and processing, producing optimal DNA yields.
After sample collection, each 2D spin-out swab is placed in an individual well of the E-Z 96® spin-out device and the unique integrated 2-D barcode on the cap allows for identification of the sample location within the 96-well plate. The need to manually input a well location and sample ID into your laboratory information management system is completely eliminated.
The E-Z 96® spin-out plate adapter’s unique design allows for samples to be raised and lowered into the lysis buffer. When the adaptors are in the lowered position, the samples are submerged in the lysis buffer, allowing for the incubation and lysis of the sample. The adapter is then raised and locked into place, which raises the baskets above the liquid level so they can be centrifuged to recover the residual liquid from the swabs. After centrifugation, the samples can be easily removed and discarded along with the spin-out adapter.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
The main product you are looking for is no longer available for purchase. Accompanying products and services may be found elsewhere on the page.
COMPLEMENTARY PRODUCTS
Expand All / Collapse All
Spin-out baskets and adapters
Description Pk VWR Catalog Number Quantity
E-Z 96® Spin-Out baskets, 10×100 1.000 AC7099
E-Z 96® Spin-Out plate adapters, 10×96 960 AC7088
E-Z 96® Spin-Out combination pack, containing 1× E-Z 96® Spin-Out adapter and 100× E-Z 96® Spin-Out baskets 1 AC7098
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
WRITE A REVIEW
Need help?
  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.
VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.
Please confirm the industry you work in
Example: Pharmaceutical
Please confirm your role
Example: Clinical Trial Assistant
Please confirm your location
Example: New York, NY
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty