HPLC columns, Avantor® ACE® HILIC-N, microbore
Supplier: Avantor®
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ACE®
HILN-5-25005SEA 644 GBP
HILN-5-25005S HILN-3-0301 HILN-3-7501S HILN-5-15005S HILN-3-0501S HILN-5-12005S HILN-3-05005S HILN-5-7501 HILN-3-7501 HILN-5-15005 HILN-5-75005 HILN-5-0301S HILN-3-03005S HILN-5-1201 HILN-5-0301 HILN-3-12005S HILN-3-10005S HILN-5-12005 HILN-3-1501S HILN-3-0301S HILN-3-0501 HILN-5-1501 HILN-5-05005S HILN-3-75005S HILN-5-2501 HILN-5-10005S HILN-5-1201S HILN-3-03005 HILN-5-0501 HILN-5-03005S HILN-5-1501S HILN-3-10005 HILN-3-1001 HILN-5-75005S HILN-3-15005S HILN-5-1001S HILN-3-05005 HILN-3-12005 HILN-5-25005 HILN-3-15005 HILN-5-03005 HILN-5-1001 HILN-5-10005 HILN-3-1501 HILN-3-1001S HILN-3-1201S HILN-5-0501S HILN-3-75005 HILN-5-05005 HILN-3-1201 HILN-5-7501S HILN-5-2501S
HPLC columns, Avantor® ACE® HILIC-N, microbore
Chromatography Columns
Avantor® ACE® HILIC-N columns have been designed specifically for HILIC applications and are especially useful for the separation of polar analytes. These columns are part of a suit of 3 proprietary HILIC columns (HILIC-A, HILIC-B and HILIC-N) developed to enable chromatographers to achieve reproducible and successful HILIC separations using a systematic approach to method development.
  • Proprietary polyhydroxy neutral character HILIC phase
  • Compatible with all manufacturers’ HPLC/UHPLC systems
  • Excellent column performance, inertness and robustness
  • Simplified and structured approach to successful HILIC separations
  • Available in a range of particle sizes and column dimensions including UHPLC/HPLC rated hardware formats
Ordering information: HILIC method development kits, HILIC method development guide and flowchart are available on request.