Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PDE11A Rabbit Polyclonal Antibody

Supplier: ProSci Inc.
Phosphodiesterase 11A is a cAMP/cGMP dual-specific phosphodiesterase. PDE11A activity is inhibited by IBMX, as well as zaprinast and dipyridamole. At least four isoforms have been identified which carry distinct GAF sequences in the N-terminal regions. PDE11A2 contains an additional 86 amino acids at the N-terminus. PDE11A3 contains an additional 108 N-terminal amino acids compared to PDE11A2. PDE11A4 is the longest isoform.

PDE11A antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 μg/mL, and immunofluorescence starting at 10 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PDE11A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bat, Bovine, Chicken, Gibbon, Gorilla, Hamster, Horse, Human, Monkey, Mouse, Pig, Rabbit, Rat, Zebrafish




 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
50 µg 50 µG PRSI48-334
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol PDE11A
Antigen name Phosphodiesterase 11A
Reactivity Chicken, Gibbon, Human, Rat, Bovine, Zebrafish, Hamster, Bat, Monkey, Horse, Pig, Rabbit, Mouse, Gorilla
Conjugation Unconjugated
Clonality Polyclonal
Host Rabbit
ImmunoChemistry Yes
Antigen synonyms dual 3',5'-cyclic-AMP and -GMP phosphodiesterase 11A, MGC133355, PDE11A, FLJ23693, MGC133356, PPNAD2 , cyclic nucleotide phosphodiesterase 11A, cAMP and cGMP cyclic nucleotide phosphodiesterase 11A, phosphodiesterase 11A
Immunogen PDE11A antibody was raised against a peptide located near the internal domain of PDE11A (Human).
Form Liquid
Tested applications Immunoaffinity Chromatography

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty