Anti-FLT3 Rabbit Polyclonal Antibody

Supplier: ProSci
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PRSI48-319EA 603 GBP
PRSI48-319
Anti-FLT3 Rabbit Polyclonal Antibody
Antibodies
FLT3, a PDGF Receptor-type protein kinase, promotes the differentiation and proliferation of hematopoietic cells by binding to hematopoietic growth factor. Mutations of the FLT3 gene have been implicated in acute myeloid leukemia.

FLT3 antibody can be used in immunohistochemistry starting at 13 - 26 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FLT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Gorilla, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR